Learn More
Lenz Laborglasintrumente Ball Members Inch Range
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
91.00€ - 134.00€
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10050341
|
Lenz Laborglasintrumente
01211909 |
S 19 |
94.00€
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10490123
|
Lenz Laborglasintrumente
01213520 |
S 35 |
134.00€
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10129650
|
Lenz Laborglasintrumente
01211305 |
S 13/5 |
91.00€
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10614661
|
Lenz Laborglasintrumente
01212915 |
S 29 |
125.00€
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
Description
- Ball members
- Inch range
- Acc. to DIN 12264
- Made of DURAN™ tubing
Specifications
Duran Glass |