Learn More
Speirs Robertson™ Balance Platform
Ensures that vibrations common in most laboratories are effectively removed, greatly increasing instrument performance. Speirs Robertson™ Balance Platform is constructed of stainless steel with damped sorbothane for vibration reduction.
Brand: Speirs Robertson™ AMS 30x56 FR
Description
- Unique Sorbothane isolator design removes up to 98 percent of unwanted vibrations
- Custom Damped Sorbothane low frequency isolators built in
- Pharma grade stainless meets stringent cleanliness requirements and is resistant to almost all forms of chemical attack
- Sound deadened layer construction eliminates surface vibrations
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.