missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker Agar Base (Dehydrated)

Product Code. p-200112228
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10147992

Brand: Thermo Scientific™ CM0275B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and enumerate Staphylococcus aureus with this selective and sensitive medium.

Isolate and enumerate Staphylococcus aureus with selective and sensitive Thermo Scientific™ Baird-Parker Agar Base (Dehydrated).

Baird Parker Agar Base contains carefully balanced selective inhibitory agents, glycine, lithium, and tellurite, to suppress the growth of most of the bacteria present in foods without inhibiting coagulase-positive staphylococci. The presence of egg yolk emulsion and sodium pyruvate aid the recovery of damaged cells for presumptive identification of Staphylococcus aureus in food specimens.

Baird-Parker Agar Base is widely recommended by national and international bodies for the isolation of Staphylococcus aureus1.

  • Easy interpretation of test results as Staphylococcus aureus forms grey-black shiny colonies with surrounding clear zones.
  • Greater recovery of damaged staphylococci than on other media2.

Baird-Parker2 developed this medium from the tellurite-glycine formulation of Zebovitz et al.3 and improved its reliability in isolating Staphylococcus aureus from foods. Baird-Parker added sodium pyruvate, to protect damaged cells and aid their recovery3

Egg yolk emulsion makes the medium yellow and opaque. Staphylococcus aureus reduces tellurite to form grey-black shiny colonies and then produces clear zones around the colonies by proteolytic action. This clear zone with typical grey-black colony is diagnostic for Staphylococcus aureus. On further incubation, most strains of Staphylococcus aureus form opaque haloes around the colonies.

Some strains of Staphylococcus saprophyticus may also produce both clear zones and opaque haloes but can be distinguished by the longer incubation required4.

Colonies typical of Staphylococcus aureus, but without an egg yolk reaction, should also be tested for coagulase production5. Egg yolk reaction negative Staphylococcus aureus may occur in some food, especially cheese.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, EP, IDF, ISO, NMKL, USDA.
Description Baird-Parker Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.9L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.