Learn More
Kartell™ L-Shaped Spreader
Kartell™ L-Shaped Spreader is great for dispersing liquids across agar culture plates and features a 100° angle for easier use.
Brand: Kartell™ 0264800
461.70 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 3.40200kg
Description
- Disposable
- Sterile PS spreaders for bacteriology with special 100° angling for easier use
- Especially handy for dispersing liquids across agar culture plates
Specifications
L-Shape | |
5 pcs. | |
CE/IVD |
White | |
Polystyrene |