Learn More
Brady™ B33 Series Polyamide-coated Nylon Cloth Labels
Nylon cloth (B-499) wrap-around labels are highly pliable and conform to tightly curved surfaces such as wire and cable.
203.00€ - 544.00€
Specifications
Autoclavable Autoclavable
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Format Die-cut
View More Specs
Products 10
Product Code Brand For Use With (Application) Height (English) Width (English) Price Quantity & Availability  
Product Code Brand For Use With (Application) Height (English) Width (English) Price Quantity & Availability  
30041416
View Documents
Brady™
153088
Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling 0.75 in. 0.75 in.
203.00€
Pack of 1000
30041434
View Documents
Brady™
153087
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 0.75 in. 1 in.
306.00€
Pack of 2500
30041418
View Documents
Brady™
153089
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 0.937 in. 0.75 in.
286.00€
Pack of 2500
30041463
View Documents
Brady™
153091
Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling 1.437 in. 0.25 in.
240.00€
Pack of 2500
30041460
View Documents
Brady™
153090
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 1.437 in. 0.5 in.
293.00€
Pack of 2500
30041481
View Documents
Brady™
153092
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 1.437 in. 1 in.
544.00€
Pack of 2300
30041443
View Documents
Brady™
153093
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 2.5 in. 1 in.
285.00€
Pack of 700
30041409
View Documents
Brady™
153094
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 4 in. 1 in.
294.00€
Pack of 450
30041426
View Documents
Brady™
153095
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 4 in. 1.5 in.
440.00€
Pack of 450
30041444
View Documents
Brady™
153096
Wire and Cable Labeling, Electrical Labeling, Data and Telecommunications Labeling, Laboratory Labeling, General Identification 6 in. 1.5 in.
440.00€
Pack of 300
Description
Description
Nylon cloth (B-499) wrap-around labels are highly pliable and conform to tightly curved surfaces such as wire and cable. A ribbon is required to print this material.
  • Matte white, polyamide-coated nylon cloth has permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • UL approved, CSA accepted
  • Wide service temperature range
Specifications
Specifications
Autoclavable
White
Die-cut
Single-side Printable
Non-sterile
-40°F to 194°F
Permanent Acrylic
Matte
Nylon Cloth
Thermal Transfer
Brady™ Label Printers
0.0065 in.