missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ B30 Series Pre-Printed GHS CLP Chemical Labels
Pre-printed GHS/CLP chemical labels for BBP3X/S3XXX/i3300 printers.
146.00€ - 217.00€
Specifications
Certifications/Compliance | BS 5609:1986 Section 2 and section 3 |
---|---|
Description | Pre-printed GHS/CLP Chemical Labels for BBP3X/S3XXX/i3300 Printers |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | Red on White |
Product Code | Brand | Display Text | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Dimensions (LxW) | Price | Quantity & Availability | ||||
30041474
|
Brady™
121642 |
3 Red Diamonds per Sticker | 110.00 mm (W) x 75.00 mm (H) |
217.00€
Pack of 270 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30041405
|
Brady™
121643 |
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker | 110.00 mm (W) x 75.00 mm (H) |
217.00€
Pack of 270 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30041456
|
Brady™
121636 |
3 Red Diamonds per Sticker | 84.99 mm (W) x 55.00 mm (H) |
146.00€
Pack of 340 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For BBP3X/S3XXX/i3300 printers.
Specifications
BS 5609:1986 Section 2 and section 3 | |
Permanent Acrylic | |
Red on White | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
BBP11, BBP12, PR Plus, THT 25 mm Core Printers | |
-20°C | |
Rectangle | |
-20°C to +75°C | |
0.102 mm |
Pre-printed GHS/CLP Chemical Labels for BBP3X/S3XXX/i3300 Printers | |
None | |
Gloss | |
Front | |
Labels | |
Facility Identification, Hazard Communication Identification, Electrical Labelling, Safety Identification | |
1 | |
B30-R10000 | |
-4°F to 167°F | |
0.004 in. |