missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-HT 933 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective α2-adrenoceptor agonist. Exhibits greater than 300-fold selectivity for the α2-adrenoceptor over the α1-adrenoceptor.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 254.16 |
Percent Purity | >99% |
Chemical Name or Material | B-HT 933 dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'