missing translation for 'onlineSavingsMsg'
Learn More

B-HT 933 dihydrochloride, Tocris Bioscience™

Product Code. 18014212
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective α2 agonist

Selective α2-adrenoceptor agonist. Exhibits greater than 300-fold selectivity for the α2-adrenoceptor over the α1-adrenoceptor.

Chemical Identifiers

CAS 36067-72-8
Molecular Formula C9H17Cl2N3O
Molecular Weight (g/mol) 254.16
MDL Number MFCD01529923
InChI Key HBLPYIOKPJVFQW-UHFFFAOYSA-N
Synonym b-ht 933 dihydrochloride, azepexole hydrochloride, azepexole dihydrochloride, 2-amino-6-ethyl-4,5,7,8-tetrahydro-6h-oxazolo 5,4-d azepine dihydrochloride, 5,6,7,8-tetrahydro-6-ethyl-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, 6-ethyl-5,6,7,8-tetrahydro-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, dsstox_cid_25756, dsstox_rid_81099, dsstox_gsid_45756, azepexole hcl
PubChem CID 169743
IUPAC Name dihydrogen 6-ethyl-4H,5H,6H,7H,8H-[1,3]oxazolo[4,5-d]azepin-2-amine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CCN1CCC2=C(CC1)N=C(N)O2

Specifications

Quantity 10 mg
Formula Weight 254.16
Percent Purity >99%
Chemical Name or Material B-HT 933 dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'