missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B-7610 Paper Hazardous Substance Labels, 25 mm Core

Product Code. 16647763
Click to view available options
Height (Metric):
85 mm
110 mm
Width (Metric):
55 mm
75 mm
Vertical Repeat (Metric):
91 mm
115.4 mm
Unit Size:
1 roll
This item is not returnable. View return policy

Product Code. 16647763

Brand: Brady™ 622330

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pre-printed, desktop printer labels for CLP-GHS hazardous substances.

  • Supplied with pre-printed red diamonds
  • Ready for you to print the required warning, compliant to the latest regulations

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Matte
Format Die-cut
Material Paper (B-7610)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) BBP™ 11, BBP™ 12, and BradyPrinter PR Plus Series Label Printers, THT 76 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 300 Labels/Roll
Quantity 300 Labels
Shape Rectangle
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Vertical Repeat (Metric) 115.4 mm
Dimensions (LxW) 110 x 75 mm
Length (English) 4.33 in.
Width (English) 2.95 in.
Width (Metric) 75 mm
Height (English) 4.33 in.
Height (Metric) 110 mm
Alternate Part Number THTCLP-M-L4B-7610-0.3-SC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.