Learn More
Azlon™ PMP Autoclavable Tweezer
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
Specifications
Tip Style | Sharp |
---|---|
Material | PMP |
Product Type | Tweezers |
For Use With (Application) | For secure handling of samples |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12054618
|
Azlon™
|
11.5 cm |
N/A
|
N/A
![]() |
|||||
12084588
|
Azlon™
|
14.5 cm |
N/A
|
N/A
![]() |
|||||
Description
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
- High quality, autoclavable tweezers
- Sharp tip
- Fine tooth grips for secure handling of samples
Specifications
Sharp | |
PMP | |
Tweezers | |
For secure handling of samples |