Learn More
Azlon™ Square Shaped HDPE Bucket
Product Code. p-8001780
Click to view available options
Color:
Black
White
Unit Size:
Each
Product Code. 12035562
Brand: Azlon BWX108
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Square shaped buckets suitable for food contact applications in accordance with EU regulations
  • Durable square shape allows economical use of space
  • Handle grip for easy lifting and carrying
  • Moulded graduations (14L x 0.5 divisions)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Square Bucket
Material HDPE
Autoclavable Not Autoclavable
Color Black
Handle Material Polyethylene
Height (Metric) 292 mm
Length (Metric) 292 mm
Quantity 1 Ea.
Width (Metric) 292 mm