missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 4547

Potent and selective FGFR inhibitor

247.00€ - 958.00€

Specifications

Chemical Name or Material rel-N-[5-[2-(3,5-Dimethoxyphenyl)ethyl]-1H-pyrazol-3-yl]-4-[(3R,5S)-3,5-dimethyl-1-piperazinyl]benzamide
CAS 1035270-39-3
Target FGF Receptor Inhibitors
Molecular Formula C26H33N5O3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18717376
View Documents Promotion Details
Tocris Bioscience™
7823/10
10 mg
247.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18718386
View Documents Promotion Details
Tocris Bioscience™
7823/50
50 mg
958.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AZD 4547 is a potent and selective FGFR1, FGFR3 and FGFR2 tyrosine kinase inhibitor (IC50 values are 0.2 1.8, and 2.5 nM, respectively). It shows selectivity for FGFR across a range of unrelated tyrosine and serine/threonine kinases, including IGFR (>2,900-fold), CDK2 (>50,000-fold), and p38 (>50,000-fold). In vitro, it has antiproliferative effects on tumor cell lines with deregulated FGFR expression. In vivo, AZD 4547 exhibits dose-dependent inhibition of tumor growth in an FGFR-driven human tumor xenograft mice model. It suppresses osteoclastogenesis and tumor-induced osteolysis in an orthotopic breast cancer bone metastasis mouse model. AZD 4547 is orally bioavailable.
Specifications

Specifications

rel-N-[5-[2-(3,5-Dimethoxyphenyl)ethyl]-1H-pyrazol-3-yl]-4-[(3R,5S)-3,5-dimethyl-1-piperazinyl]benzamide
1035270-39-3
FGF Receptor Inhibitors
C26H33N5O3
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documents

Documents

Product Certifications