Learn More
Axygen™ Axypet™ Single Channel Pipettor, Calibration 3x10 STD
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
252.27€ - 278.42€
Specifications
Certifications/Compliance ISO17025
Number of Channels 1
Product Line Axypet™
Type Single Channel Pipettor
Quantity 1/Pk., 1/Cs
View More Specs
Products 10
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17203624
View Documents
Axygen™
AP-2-STD
0.1 to 2 μL
252.27€
Each
In Stock
to see stock.
Add to basket
17243604
View Documents
Axygen™
AP-10-STD
0.5 to 10 μL
252.27€
Each
17203634
View Documents
Axygen™
AP-5000-STD
1 to 5 mL
278.42€
Each
17203614
View Documents
Axygen™
AP-10ML-STD
1 to 10 mL
278.42€
Each
Add to basket
17223624
View Documents
Axygen™
AP-20-STD
2 to 20 μL
252.27€
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
17283624
View Documents
Axygen™
AP-50-STD
5 to 50 μL
252.27€
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
17263604
View Documents
Axygen™
AP-100-STD
10 to 100 μL
252.27€
Each
17243624
View Documents
Axygen™
AP-200-STD
20 to 200 μL
252.27€
Each
17263624
View Documents
Axygen™
AP-250-STD
50 to 250 μL
252.27€
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
17283604
View Documents
Axygen™
AP-1000-STD
100 to 1000 μL
252.27€
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
Description
Description
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement (Left handed EZIject button available by special order).
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications
Specifications
ISO17025
Axypet™
1/Pk., 1/Cs
1
Single Channel Pipettor