missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axl Antibody (enapotamab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28634JF635
This item is not returnable.
View return policy
Description
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Axl | |
| Monoclonal | |
| Janelia Fluor 635 | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| AXL/ UFO | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 558 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| enapotamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction