missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Avermectin, ACROS Organics™

119.60€ - 394.97€

Specifications

Chemical Name or Material Avermectin
CAS 65195-55-3
Molecular Formula C48 H72 O14
Formula Weight 873.09
Physical Form Crystalline Powder
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17362481
View Documents
Thermo Scientific™
468392500
250 mg
119.60€
250mg
Please to purchase this item. Need a web account? Register with us today!
17352481
View Documents
Thermo Scientific™
468390010
1 g
394.97€
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Avermectin
C48 H72 O14
Crystalline Powder
65195-55-3
873.09
White to Light-yellow
Videos
Safety and Handling

Safety and Handling

GHS08: Health hazard

GHS09: Environment

GHS06: Skull and crossbones

missing translation for 'einecsNumber' : 265-610-3

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Product Certifications