Learn More
Abeba™ Autoclavable Clogs 9620
Antistatic and autoclavable at 121°C. High slip resistance.
61.65€ - 63.55€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRC |
---|---|
Color | Green |
Style | 9620 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15673258
|
Abeba™
9620/36 |
36 |
61.65€
1 pair |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
15683258
|
Abeba™
9620/37 |
37 |
63.55€
1 pair |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15693258
|
Abeba™
9620/38 |
38 |
63.55€
1 pair |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
15603268
|
Abeba™
9620/39 |
39 |
63.55€
1 pair |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15613268
|
Abeba™
9620/40 |
40 |
63.55€
1 pair |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15623268
|
Abeba™
9620/41 |
41 |
63.55€
1 pair |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15633268
|
Abeba™
9620/42 |
42 |
63.55€
1 pair |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
15643268
|
Abeba™
9620/43 |
43 |
61.65€
1 pair |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15653268
|
Abeba™
9620/44 |
44 |
61.65€
1 pair |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
15663268
|
Abeba™
9620/45 |
45 |
63.55€
1 pair |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15673268
|
Abeba™
9620/46 |
46 |
61.65€
1 pair |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- Green
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
Green | |
9620 |