Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand: 3M™ 7000034739
This item is not returnable.
View return policy
Description
Fits a wide range of face shapes and sizes- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions