Learn More
3M™ Aura™ Particulate Respirator
Particulate respirator with 3-panel design allows for greater facial movement
Brand:  3M™ 7000088723
 View more versions of this product
Product Code. 17217620
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Original 3-panel design allows for greater facial movement
  • Low-resistance filter technology helps you breathe more easily
  • Sculpted upper panel conforms to the nose and contours of the face for an effective seal
  • Embossed top panel helps reduce eyewear fogging
  • Chin tab designed for ease of positioning and adjustment
  • Easy to store away with flat-fold design and individual wrapping
  • Colour-coded headbands for easy identification: yellow for FFP1 (NFP 4)
  • Sculpted nose panel helps conform to the nose and contours of the face and helps to improve compatibility with 3M eyewear
  • Maximum usage level: Up to 4 x TLV
  • 3M™ Cool Flow™ Valve helps reduce heat build-up in hot and humid conditions
  • Available in large or small pack sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FFP2
9320+
Particulate Respirator
No