Learn More
3M™ Aura™ FFP1-FFP3 Particulate Respirators
3M™ Aura™ Particulate Respirator provides breathing protection with a flat-fold, 3-panel design, low breathing resistance and an embossed, sculpted top panel that can help reduce eyewear fogging.
Brand:  3M™ 7000088812
 View more versions of this product
Product Code. 17257620
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Providing a unique three-panel design for greater comfort and easy communication. 3M™ Aura™ 9300 Series Particulate Respirators cover a range of applications requiring FFP1, FFP2 and FFP3 protection.
  • Individually packaged to avoid contamination
  • Soft cover web on inner panel ensures greater comfort
  • Lightweight and foldable
  • Evenly tensioned headbands to relieve strap pressure
  • Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Approved to EN 149:2001A1:2009
3-panel
FFP3
No
Yes
Sculpted
Cup
White
Single-use
Flat fold Particulate Respirator
Agriculture, Cleaning Maintenance, Demolition
9330
Dusts and mists found in a wide variety of industrial applications and other work situations