Learn More
3M™ Aura™ 9300 Series Particulate Respirators
Providing a unique three-panel design for greater comfort and easy communication. 3M™ Aura™ 9300 Series Particulate Respirators cover a range of applications requiring FFP1, FFP2 and FFP3 protection.
Brand:  3M™ 7000088812
Product Code. 11757574
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Individually packaged to avoid contamination
  • Soft cover web on inner panel ensures greater comfort
  • Lightweight and foldable
  • Evenly tensioned headbands to relieve strap pressure
  • Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN149: 2001 A1: 2009 FFP3 NR D
3-panel
Dust Mask
General industry, cement dust, pharmaceuticals, pottery and ceramics, career exploitation
20/Pk.
Pliable
Red
FFP3
Yes
9330 Dust Mask
Fine dust, water and oil mist