Learn More
Attune™ NxT Software, single license
Attune NxT Software, single license
POA
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15320677
View Documents
Invitrogen
A25554
1 license
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15340667
View Documents
Invitrogen
A24855
10 Licenses
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15350667
View Documents
Invitrogen
A24856
5 Each
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Attune™ NxT Software is used to operate and maintain the Attune™ NxT acoustic focusing cytometer. It enables powerful user-defined experimental analysis via an intuitive, easy-to-use interface and provides advanced tools for both acquisition and analysis that are simple enough for users at all experience levels.
  • Simplified use—streamlined, intuitive interface
  • Speed—faster real-time analysis of large data sets
  • Easy instrument operation—automated maintenance and instrument performance monitoring
    Simplified use
    Attune™ NxT software employs a menu system that presents the features you need when you need them without a lot of scrolling and clicking. The customization tab allows you to pre-define settings for plots, gates, experiments, reports, and overlays. To simplify experimental set up, the preview screen shows all plot combinations instantaneously allowing you to easily add them to the experiment workspace. Compensation is automated or user-defined and can be set up using a compensation guide.
    Speed increases productivity
    Attune™ NxT Software has been designed to maximize the efficiency in performing data analysis. It uses a Windows™ 7 64-bit system to provide fast refresh rates for large data sets of up to 20 million events created by multi-laser, multi-parameter instrumentation. The software allows you to immediately visualize changes to your data plots in real-time as you make adjustments. This allows you to spend less time analyzing data and more time on your research.
    Easy instrument operation
    Flow cytometers often require the user to spend time ensuring the quality of the system through maintenance tasks and proper QC, since changes in lasers, fluidics, and optics can affect the quality of the data generated. Attune™ NxT Software takes many of these manual tasks and fully automates them to help reduce time spent on instrument maintenance. The software provides an automated quality control instrument tracking system, important for collecting and analyzing accurate data requiring minimal user interaction.
    Software features:
    Instrument controls
    Automated maintenance and performance testing
    Data collection
    Visualization tools
    Dot plot, density plot, histogram
    Histogram marker, quad marker
    Customized logic gates
    Data plot preview for quick plot set-up
    Data display—log, linear, log transforms
    Statistics—mean, median, mode, %CV, rCV, SD, % total, % gated
    Experiment set-up
    Automated compensation
    Auto layout and freeform workspace layouts
    Analysis tools
    Multi-file selection tools
    File management
    Import/export of files and experiments
    Files stored as FCS3.1 format
    User management & security
    User and administrator accounts—basic/admin
  • Videos
    SDS
    Documents
    Documents
    Product Certifications
    For Research Use Only. Not for use in diagnostic procedures.
    Product Content Correction
    Product Title
    En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
    Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis