Learn More
WTW™ Attachment Stirrer for WTW™ Oxygen Probe
Brand:  WTW™ 203824
Product Code. 10099260
  • 191.00€ / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Attachment Stirrer
Provides a constant, defined flow of the sensor, in conjunction with the Oxi-Stirrer 300/magnetic stirrer
Oxygen probe