missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Atomatic Pipet Duran

Product Code. 10622821
Click to view available options
Capacity (Metric):
5 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • Kipp dispensers
  • Dispenser head with bottle (1 L)
  • Ground joint connection: NS 29/32

Specifications

Capacity (Metric) 100 mL
Standard Taper Joint NS 29/32
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'