missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Athletic Lite Assembly Gloves

Product Code. 17260331
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are constructed from a combination of polyamide and elastane, with a nitrile rubber microfoam coating for enhanced grip in dry or slightly damp applications.

  • Slim, ergonomic fit allows for precision work with high levels of user comfort
  • Fine-structured, open-pored microfoam coating enhances grip capabilities
  • Breathable, matte coating reduces excessive perspiration
  • Ideal for precision assembly work, maintenance, inspection, and sorting
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR MicroFoam
Color Blue, Black
Material Polyamide, Elastane
Size 7
EN388 Standard 4132X
For Use With (Application) Inspection, Precision Assembly Work, Repair Work, and Sorting
Gender Unisex
Glove Cuff Style Knit
Series Athletic Lite
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'