missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Athletic Lite Assembly Gloves
Click to view available options
Size:
7
8
9
10
11
Description
- Slim, ergonomic fit allows for precision work with high levels of user comfort
- Fine-structured, open-pored microfoam coating enhances grip capabilities
- Breathable, matte coating reduces excessive perspiration
- Ideal for precision assembly work, maintenance, inspection, and sorting
- Certified by OEKO-TEX™ Standard 100
Specifications
Specifications
Certifications/Compliance | OEKO-TEX™ Standard 100 |
Applications | Industrial |
Coating Material | NBR MicroFoam |
Color | Blue, Black |
Material | Polyamide, Elastane |
Size | 11 |
EN388 Standard | 4132X |
For Use With (Application) | Inspection, Precision Assembly Work, Repair Work, and Sorting |
Gender | Unisex |
Glove Cuff Style | Knit |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'