Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Product Code. 12460138
Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Unit Size:
Pack of 100
Product Code. 12460138
Brand: Thermo Scientific™ C401361R
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
TRUSTED_SUSTAINABILITY
Specifications
Description PP, Red; Kim-Lok Red PTFE/White Silicone septum
Color Red
Material PTFE, Silicone, Polypropylene
Septum Kim-Lok Red PTFE/White Silicone
Type Kim-Lok Cap with Flange
Closure Size 8-425
For Use With (Equipment) 8-425 Standard Opening Vials