missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ARN 14494
Potent serine palmitoyltransferase inhibitor
190.06€ - 706.25€
Specifications
Chemical Name or Material | rel-1-[1-[2-[(3 aR,6 AS)-Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl]-2-oxoethyl]-4-piperidinyl]-2,3-dihydro-N-methyl-2-oxo-1 H-indole-5-carboxamide |
---|---|
CAS | 1037834-43-7 |
Target | Other Transferase Inhibitors |
Molecular Formula | C24H32N4O3 |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718176
|
Tocris Bioscience™
7134/10 |
10 mg |
190.06€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765736
|
Tocris Bioscience™
7134/50 |
50 mg |
706.25€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARN 14494 is a potent serine palmitoyltransferase inhibitor (IC50 = 27.3 nM). ARN 14494 inhibits synthesis of long chain ceramides and dihydroceramides in an in vitro model of Alzheimer's diease. The compound prevents the synthesis of proinflammatory cytokines and the oxidative stress-related enzymes iNOS and COX-2 in mouse primary cortical astrocytes. ARN 14494 is neuroprotective against β-amyloid 1-42 induced neurotoxicity in primary cortical neurons co-cultured with astrocytes.Specifications
rel-1-[1-[2-[(3 aR,6 AS)-Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl]-2-oxoethyl]-4-piperidinyl]-2,3-dihydro-N-methyl-2-oxo-1 H-indole-5-carboxamide | |
Other Transferase Inhibitors | |
0.98 |
1037834-43-7 | |
C24H32N4O3 |
Safety and Handling
Recommended Storage : Store at -20°C