Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I (250 units/tube)
A 94 kDa, thermostable, recombinant DNA polymerase
Brand:  Applied Biosystems™ N8080166
Product Code. 10486924
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
AmpliTaq™ DNA Polymerase is recombinant DNA polymerase obtained by expression of modified form of Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is thoroughly characterized enzyme available for PCR process and its recombinant nature and purification method provide unparalleled purity and reproducibility
  • Characterized enzyme available for the PCR process
  • QC-tested to guarantee reproducible results
  • Ideal for PCR
PCR, PCR and Real-Time PCR, Routine PCR
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 x 200μL of AmpliTaq DNA Polymerase (at 5U/μL); 4 x 1.5mL of GeneAmp 10X PCR Buffer (100mM Tris-HCl pH 8.3, 500mM KCl, 15mM MgCl2,0.01% (w/v) gelatin. Store at -20°C.
5' - 3'
1 X
5 kb or less
AmpliTaq DNA Polymerase
1500 units
3'-A