missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Antibiotics: Nystatin
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Brand: MP Biomedicals™ 0219453405
View more versions of this product
This item is not returnable.
View return policy
Description
- Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
4°C | |
Powder | |
Antifungal | |
5 mU |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction