missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XMRV Virus Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532642
This item is not returnable.
View return policy
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is prostate adenocarcinoma with XMRV infection.
Xenotropic murine leukemia virus-related virus (XMRV) is a newly recognized human retrovirus first identified in prostate cancer tissues from men with a deficiency in the innate immunity gene RNASEL. At present, studies have detected XMRV DNA and protein in prostate cancer at widely different rates and in patients with chronic fatigue syndrome. The XMRV protein is synthesized in the Golgi by host cells.Specifications
XMRV Virus | |
Polyclonal | |
Xenotropic murine leukemia virus-related virus | |
Synthetic peptide near N-terminus of XMRV envelope protein. | |
500 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction