Learn More
Sartorius Anti-Static Devices for Balances
GREENER_CHOICE
Product Code. 10465765 Shop All Sartorius Products
Click to view available options
Type:
Anti-static Pen
Unit Size:
Each
Product Code. 10465765
Brand: Sartorius YSTP01
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Control charges on samples
  • Anti-static pen—direct the pen at item to be neutralized
  • Ionizing blower—point blower toward sample prior to weighing
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, SE and ME Ultramicro Balance
Type Anti-static Pen