Learn More
anti-SPINK1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00006690-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-SPINK1, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-SPINK1, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
SPINK1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SPINK1. | |
PCTT/PSTI/Spink3/TATI | |
Mouse | |
RUO | |
6690 | |
Human | |
Liquid |
ELISA | |
Biotin | |
SPINK1 | |
SPINK1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SPINK1, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SPINK1, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only