missing translation for 'onlineSavingsMsg'
Learn More

anti-MSH2, E2F3 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0360.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16131304

  • 632.00€ / 1 set
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: MSH2 rabbit purified polyclonal antibody (20μg); E2F3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MSH2 protein, and the other against the E2F3 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

MSH2
Monoclonal/Polyclonal
MSH2
MSH2
1 Set
Primary
Antibody pair set content: 1. MSH2 rabbit purified polyclonal antibody (20 ug) 2. E2F3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
COCA1/FCC1/HNPCC/HNPCC1/LCFS2
Mouse, Rabbit
RUO
4436/1871
Human
MSH2 & E2F3 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers