Learn More
anti-MAP3K14, CASP3 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0597.1Set
Additional Details : Weight : 0.00010kg
Includes: MAP3K14 rabbit purified polyclonal antibody (20μg); CASP3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP3K14 protein, and the other against the CASP3 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
MAP3K14 | |
Monoclonal/Polyclonal | |
MAP3K14 | |
MAP3K14 | |
1 Set | |
Primary | |
Antibody pair set content: 1. MAP3K14 rabbit purified polyclonal antibody (20 ug) 2. CASP3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
FTDCR1B/HS/HSNIK/NIK | |
Mouse, Rabbit | |
RUO | |
9020/836 | |
Human | |
MAP3K14 & CASP3 Protein Protein Interaction Antibody Pair |