missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-LAMA5, LAMC1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0341.1Set
Includes: LAMA5 rabbit purified polyclonal antibody (20μg); LAMC1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the LAMA5 protein, and the other against the LAMC1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
LAMA5 | |
Monoclonal/Polyclonal | |
LAMA5 | |
LAMA5 | |
1 Set | |
Primary | |
Antibody pair set content: 1. LAMA5 rabbit purified polyclonal antibody (20 ug) 2. LAMC1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
KIAA1907 | |
Mouse, Rabbit | |
RUO | |
3911/3915 | |
Human | |
LAMA5 & LAMC1 Protein Protein Interaction Antibody Pair |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto