missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Anti-Human Control γ1 FITC/γ1 PE/CD45 PerCP with BD Trucount™ Tubes (RUO/GMP)

Three-color direct immunofluorescence reagent used as an isotype (negative) control for flow cytometric immunophenotyping of erythrocyte-lysed whole blood with BD Tritest reagents on the BD family of flow cytometers.

Brand:  BD Biosciences™ 337079

Product Code. 15865818

  • 1370.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Clone 2D1 X40 X40 (RUO (GMP))
  • BD Tritest™ Control, γ1 fluorescein isothiocyanate (FITC)/γ1 phycoerythrin (PE)/CD45 peridinin chlorophyll (PerCP), is a three-color direct immunofluorescence reagent used as an isotype (negative) control for flow cytometric immunophenotyping of erythrocyte-lysed whole blood with BD Tritest reagents on the BD family of flow cytometers.
  • The BD Tritest Control is used to determine the unstained lymphocyte population on a fluorescence 1 (FL1) vs fluorescence 2 (FL2) display and to determine any nonantigen-specific antibody binding (nonspecific staining) present, particularly that caused by Fc receptors.
  • BD Tritest Control is for in vitro diagnostic use with BD in vitro diagnostic BD Tritest reagents.
  • Contents:
  • Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
  • Clone: X40, Format: FITC, Isotype: IgG<sub>1</sub> K,
  • Clone: X40, Format: PE, Isotype: IgG<sub>1</sub> K
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration.
50 Tests
Immunofluorescence Flow Cytometry Test Reagents
Trucount Tubes
RUO (GMP)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions