missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HIP1 Ab-3 Matched Antibody Pair, Abnova™
Brand: Abnova H00003092-AP51.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-HIP1 (100μg); Detection antibody: rabbit purified polyclonal anti-HIP1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended
Description
Host Species: Mouse, Rabbit
Cross Reactivity: Human
ELISA Pair
Specifications
HIP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HIP1. | |
ILWEQ/MGC126506 | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HIP1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-HIP1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
HIP1 | |
HIP1 | |
1 Set | |
3092 | |
Human |