Learn More
anti-GSTP1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00002950-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16055678
  • 789.00€ / 1 set
Estimated Shipment: 26-07-2024
to see stock.
Includes: Antibody pair for IP: mouse monoclonal anti-GSTP1 (300μg), Antibody pair for WB: rabbit polyclonal anti-GSTP1 (50μL)
Specifications
GSTP1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
DFN7/FAEES3/GST3/PI
Mouse, Rabbit
RUO
2950
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
GSTP1
GSTP1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-GSTP1 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-GSTP1 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.