Learn More
anti-FUCA1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00002517-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16025298
  • 789.00€ / 1 set
Estimated Shipment: 26-07-2024
to see stock.
Includes: Antibody pair for IP: rabbit polyclonal anti-FUCA1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-FUCA1 (50μg)
Specifications
FUCA1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FUCA
Mouse, Rabbit
RUO
2517
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
FUCA1
FUCA1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-FUCA1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-FUCA1 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.