Learn More
anti-FOXA3, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00003171-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16035918
  • 789.00€ / 1 set
Estimated Shipment: 26-07-2024
to see stock.
Includes: Antibody pair for IP: rabbit polyclonal anti-FOXA3 (300μL), Antibody pair for WB: mouse polyclonal anti-FOXA3 (50μL)
Specifications
FOXA3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FKHH3/HNF3G/MGC10179/TCF3G
Mouse, Rabbit
RUO
3171
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
FOXA3
FOXA3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-FOXA3 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-FOXA3 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.