Learn More
cytochrome P450, family 26, subfamily B, polypeptide 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CYP26B1.
Brand: Abnova H00056603-A01.50uL
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids. The enzyme encoded by this gene is involved in the specific inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid. [provided by RefSeq
Sequence: SNSIGDIHRNKRKVFSKIFSHEALESYLPKIQLVIQDTLRAWSSHPEAINVYQEAQKLTFRMAIRVLLGFSIPEEDLGHLFEVYQQFVDNVFSLPVDLPFSpecifications
| cytochrome P450, family 26, subfamily B, polypeptide 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CYP26B1. | |
| CYP26B1 | |
| CYP26A2/DKFZp686G0638/MGC129613/P450RAI-2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 56603 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_019885 | |
| CYP26B1 | |
| CYP26B1 (NP_063938, 131 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.