Learn More
anti-CD70, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00000970-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16023838
  • 789.00€ / 1 set
Estimated Shipment: 26-07-2024
to see stock.
Includes: Antibody pair for IP: rabbit polyclonal anti-CD70 (300μL), Antibody pair for WB: mouse purified polyclonal anti-CD70 (50μg)
Specifications
CD70
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CD27L/CD27LG/TNFSF7
Mouse, Rabbit
RUO
970
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
CD70
CD70
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-CD70 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-CD70 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.