missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CACYBP, CTNNB1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0611.1Set
Includes: CACYBP rabbit purified polyclonal antibody (20μg); CTNNB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
In situ PLA
Specifications
CACYBP | |
Monoclonal/Polyclonal | |
CACYBP | |
CACYBP | |
1 Set | |
27101/1499 | |
Human | |
CACYBP & CTNNB1 Protein Protein Interaction Antibody Pair |
In situ PLA | |
Unconjugated | |
GIG5/MGC87971/PNAS-107/RP1-102G20.6/S100A6BP/SIP | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. CACYBP rabbit purified polyclonal antibody (20 ug) 2. CTNNB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction