Learn More
anti-BAG5, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00009529-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16061579
  • 789.00€ / 1 set
Estimated Shipment: 29-07-2024
to see stock.
Includes: Antibody pair for IP: rabbit polyclonal anti-BAG5 (300μL), Antibody pair for WB: mouse purified polyclonal anti-BAG5 (50μg)
Specifications
BAG5
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
BAG-5
Mouse, Rabbit
RUO
9529
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
BAG5
BAG5
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-BAG5 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-BAG5 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.