missing translation for 'onlineSavingsMsg'
Learn More

anti-AXL Matched Antibody Pair, Abnova™

Brand:  Abnova H00000558-AP51.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16063258

  • 789.00€ / 1 set
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Capture antibody: mouse monoclonal anti-AXL (100μg); Detection antibody: rabbit purified polyclonal anti-AXL (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human AXL

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair (Transfected Lysate)

Specifications

Specifications

AXL
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human AXL.
JTK11/UFO
Mouse, Rabbit
RUO
558
Human
Purified
ELISA
Unconjugated
AXL
AXL
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-AXL (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-AXL (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions