missing translation for 'onlineSavingsMsg'
Learn More

Antalarmin hydrochloride, Tocris Bioscience™

Product Code. 18045972
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CRF1 antagonist

Non-peptide corticotropin-releasing hormone receptor 1 (CRF1) antagonist (Ki = 1 nM). Suppresses CRF-induced ACTH secretion. Blocks CRF- and novelty-induced anxiety-like behavior in animal models of anxiety.

Chemical Identifiers

CAS 220953-69-5
Molecular Formula C24H35ClN4
Molecular Weight (g/mol) 415.022
InChI Key CGDGXEDXEXACKQ-UHFFFAOYSA-N
Synonym antalarmin hydrochloride, n-butyl-n-ethyl-2,5,6-trimethyl-7-2,4,6-trimethylphenyl-7h-pyrrolo 2,3-d pyrimidin-4-amine hydrochloride, n-butyl-n-ethyl-2,5,6-trimethyl-7-2,4,6-trimethylphenyl-7h-pyrrolol 2,3-d pyrimidin-4-amine hydrochloride, antalarmin hydrochloride hplc , solid, n-butyl-n-ethyl-2,5,6-trimethyl-7-2,4,6-trimethylphenyl-7h-pyrrolo 2,3-d pyrimidin-4-amine-hydrogen chloride 1/1
PubChem CID 16078945
IUPAC Name N-butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)pyrrolo[2,3-d]pyrimidin-4-amine;hydrochloride
SMILES CCCCN(CC)C1=NC(=NC2=C1C(=C(N2C3=C(C=C(C=C3C)C)C)C)C)C.Cl

Specifications

Quantity 10 mg
Formula Weight 415.01
Percent Purity >99%
Chemical Name or Material Antalarmin hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'