missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anillin Monoclonal Antibody (KAA135_9A4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556076
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TPNTKAIQER
This gene encodes an actin-binding protein that plays a role in cell growth and migration, and in cytokinesis. The encoded protein is thought to regulate actin cytoskeletal dynamics in podocytes, components of the glomerulus. Mutations in this gene are associated with focal segmental glomerulosclerosis 8. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Anillin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9NQW6 | |
| ANLN | |
| Human ANLN peptide (immunogen range is 401-410). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| KAA135_9A4 | |
| Unconjugated | |
| ANLN | |
| 1110037A17Rik; 2900037I21Rik; actin-binding protein anillin; Anillin; anillin actin binding protein; anillin, actin binding protein; anillin, actin binding protein (scraps homolog, Drosophila); Anln; C78101; DKFZp779A055; FSGS8; RGD1561367; scra; Scraps | |
| Mouse | |
| Protein A | |
| RUO | |
| 54443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction