missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Angiotensin I (human, mouse, rat)

Endogenous precursor to angiotensin II (Cat. No. 1158)

Brand:  Tocris Bioscience™ 1563/1

Product Code. 18013862

  • 67.00€ / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Angiotensin I (human, mouse, rat) is an endogenous peptide substrate for angiotensin converting enzyme (ACE) and ACE2; precursor to the vasoconstrictor peptide angiotensin II (Cat. No. 1158).
Specifications

Specifications

Human, Mouse, Rat
Store at -20°C
M.W. (Observed): 1296.5
1 mg
0.95
Angiotensin I (human, mouse, rat)
Angiotensin I (human, mouse, rat)
Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only