Learn More
Fisherbrand™ Anemometer Helix
Brand: Fisherbrand™ AM4206M
This item is not returnable.
View return policy
Specifications
Anemometer Helix | |
0.5 to 40 m/sec. | |
72 mm | |
±2% | |
-10 to +50°C |
±2% | |
180 mm | |
9V battery | |
m/s, ft/min, km/h, knots and mile/h | |
700 g |