Learn More
Agilent BioTek AMX Automated Media Exchange
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
MFXP2, MFXP2R
Unit Size:
Each
Description
- AMX Automated Media Exchange module protects 3D cell structures (e.g. spheroids, tumoroids), and suspension cells
- For use with Agilent BioTek Microplate readers and imagers
Specifications
Specifications
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Description | AMX Automated Media Exhange module |
| For Use With (Application) | Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization |
| For Use With (Equipment) | MFXP2, MFXP2R |
| Includes | AMX Autmated Media Exchange module, second peri pump, AMX dispense and aspirate manifolds |